Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taco1os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019365.1 | - | 1 | 12 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712800.1 | - | 1 | 12 | Chr. 11 - 106065080 - 106065932 [-] on Build GRCm38 |