Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038203.1 | - | 201 | |||
AK147232.1 | - | 5606 | ||||
AK153220.1 | - | 1941 |