Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other 5430414B12RIK SIRNA Assays ›
Gene Symbol:
5430414B12Rik
Gene Name:
RIKEN cDNA 5430414B12 gene
Gene Aliases:
-
Chromosome Location:
-
UniGene ID:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
n425833
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: 5430414B12Rik
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
AK013463.1
-
1
287
AK017306.1
-
1
81
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
Transcript Location
GenBankmRNA
BY717118.1
-
1
79
Chr. 1 - 191817600 - 191819429 [+] on Build GRCm38
GenBankmRNA
BF146919.1
-
1
181
Chr. 1 - 191817512 - 191818811 [+] on Build GRCm38
GenBankmRNA
BE198858.1
-
1
242
Chr. 1 - 191817437 - 191818918 [+] on Build GRCm38
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9bfc7:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline