Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9030205G03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020245.1 | - | 1 | 347 |
Gene Symbol: Dlgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK082945.1 | - | 1 | 84 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720642.1 | - | 1 | 347 | Chr. 8 - 14381732 - 14507248 [+] on Build GRCm38 |