Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700061J23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006857.1 | - | 3 | 351 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706844.1 | - | 3 | 352 | Chr. 11 - 23152140 - 23166377 [+] on Build GRCm38 |