Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm15545 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041357.1 | - | 2 | 1677 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA717989.1 | - | 1 | 260 | Chr. 7 - 44989913 - 44990389 [+] on Build GRCm38 |