Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102640923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK158984.1 | - | 6 | 620 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY646700.1 | - | 3 | 191 | Chr. 8 - 76605226 - 76797032 [-] on Build GRCm38 |