Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn34c4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015117.1 | - | 1, 2 | 129 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714663.1 | - | 1, 2 | 129 | Chr. X - 127721199 - 127736554 [-] on Build GRCm38 |