Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lincenc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110430.1 | 3 | 431 | |||
NR_110431.1 | 2 | 347 | ||||
NR_110432.1 | 2 | 393 | ||||
GenBank mRNA | BC065997.1 | - | 2 | 393 | ||
BY736053.1 | - | 3 | 432 |