Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080597.1 | - | 1 | 1440 | ||
BC059899.1 | - | 1 | 1650 | |||
BC096016.1 | - | 1 | 1650 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB264132.1 | - | 1 | 51 | Chr. 2 - 181078367 - 181078628 [-] on Build GRCm38 |