Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080597.1 | - | 1 | 313 | ||
BC059899.1 | - | 1 | 523 | |||
BC096016.1 | - | 1 | 523 | |||
CA325321.1 | - | 1 | 523 |