Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080597.1 | - | 1 | 1371 | ||
BC059899.1 | - | 1 | 1581 | |||
BC096016.1 | - | 1 | 1581 |