Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700030F04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045731.1 | 2 | 153 | |||
GenBank mRNA | AK006540.1 | - | 2 | 154 | ||
BY706558.1 | - | 2 | 154 |
Gene Symbol: 1700030N03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045304.1 | 2 | 158 | |||
GenBank mRNA | AK006563.1 | - | 154 | |||
AK007025.1 | - | 2 | 159 |
Gene Symbol: 4930533I22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029757.1 | - | 6 | 422 |
Gene Symbol: Gm38527 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077035.1 | - | 170 |
Gene Symbol: Herc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015214.1 | - | 153 |