Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700030F04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045731.1 | 2 | 127 | |||
GenBank mRNA | AK006540.1 | - | 2 | 128 | ||
BY706558.1 | - | 2 | 128 |
Gene Symbol: 1700095A21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045468.1 | 2 | 128 | |||
GenBank mRNA | AK007074.1 | - | 2 | 129 | ||
AK007118.1 | - | 2 | 130 | |||
AK161354.1 | - | 153 |
Gene Symbol: 4930533I22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029757.1 | - | 6 | 396 |
Gene Symbol: 4933438K21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045446.1 | 2 | 127 | |||
GenBank mRNA | AK017111.1 | - | 2 | 128 |
Gene Symbol: Herc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015214.1 | - | 127 |