Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700102J08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007112.1 | - | 971 | |||
AK020344.1 | - | 2 | 173 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721154.1 | - | 2 | 173 | Chr. 7 - 129918790 - 129920269 [-] on Build GRCm38 |