Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9330188P03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102319.1 | 2 | 526 | |||
GenBank mRNA | AK034417.1 | - | 1 | 3036 | ||
AK145081.1 | - | 2063 | ||||
FJ744746.1 | - | 2 | 448 | |||
FJ860514.1 | - | 2 | 448 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF101283.1 | - | 1 | 240 | Chr. 14 - 105592853 - 105593911 [+] on Build GRCm38 |