Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnd3os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040759.1 | 2 | 504 | |||
GenBank mRNA | AK018929.1 | - | 158 | |||
AK131816.1 | - | 2 | 504 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707553.1 | - | 2 | 499 | Chr. 3 - 105451270 - 105452950 [-] on Build GRCm38 |