Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9430022A07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020432.1 | - | 1 | 760 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB103512.1 | - | 1 | 83 | Chr. 10 - 116463853 - 116464138 [-] on Build GRCm38 |