Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm12153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090039.1 | - | 2 | 460 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY733084.1 | - | 2 | 456 | Chr. 11 - 43959089 - 44005189 [+] on Build GRCm38 |