Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam198b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132613.1 | - | 680 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV250422.2 | - | 1 | 374 | Chr. 3 - 79859295 - 79886500 [-] on Build GRCm38 |