Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm20742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076995.1 | - | 1 | 2971 |
Gene Symbol: Stk-ps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037690.1 | 2 | 1813 | |||
NR_037691.1 | 3 | 1933 | ||||
GenBank mRNA | AK029722.1 | - | 4 | 2775 | ||
BC145241.1 | - | 2 | 1813 | 9053793 | ||
BC145242.1 | - | 3 | 1933 | 9053794 | ||
BC150733.1 | - | 3 | 1933 | 9087644 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV271855.2 | - | 1 | 148 | Chr. 17 - 66196188 - 66199820 [+] on Build GRCm38 |