Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt6os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028111.1 | - | 318 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB610333.1 | - | 3 | 318 | Chr. 15 - 100695780 - 100704206 [+] on Build GRCm38 |