Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079567.1 | - | 2400 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB180842.2 | - | 1 | 487 | Chr. 10 - 42180224 - 42180779 [-] on Build GRCm38 |