Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009947.1 | - | 1 | 852 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN458386.1 | - | 1 | 65 | Chr. 19 - 21282901 - 21283643 [+] on Build GRCm38 |