Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm13481 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042510.1 | - | 330 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB639467.1 | - | 3 | 330 | Chr. 2 - 48063646 - 48457245 [-] on Build GRCm38 |