Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028852.1 | - | 1 | 2304 | ||
AK045169.1 | - | 1 | 13 | |||
AK050825.1 | - | 1 | 150 |