Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930549G23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045376.1 | 4 | 456 | |||
GenBank mRNA | AK016077.1 | - | 4 | 458 |