Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2210039B01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_044985.1 | 4 | 866 | |||
GenBank mRNA | AK008780.1 | - | 4 | 872 |