Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4833419F23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040328.1 | 4 | 700 | |||
GenBank mRNA | AK014726.1 | - | 4 | 702 |