Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4833419F23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040328.1 | 1 | 260 | |||
GenBank mRNA | AK014726.1 | - | 1 | 262 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AI316660.1 | - | 1 | 216 | Chr. 18 - 4353591 - 4368671 [+] on Build GRCm38 |