Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9030411K21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI552043.1 | - | 209 | |||
AK018501.1 | - | 960 |
Gene Symbol: 4930520O04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC020150.1 | - | 2182 |