Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1500037F05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005366.1 | - | 1 | 781 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY705536.1 | - | 1 | 789 | Chr. 7 - 27670918 - 27671789 [-] on Build GRCm38 |