Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCHLAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104319.1 | 3 | 505 | |||
NR_104320.1 | 3 | 505 | ||||
NR_104322.1 | 5 | 744 | ||||
NR_104323.1 | 4 | 643 | ||||
NR_104324.1 | 4 | 606 | ||||
NR_104325.1 | 4 | 606 | ||||
GenBank mRNA | JX117418.1 | - | 3 | 505 | ||
JX117419.1 | - | 3 | 505 | |||
JX117421.1 | - | 5 | 744 | |||
JX117422.1 | - | 4 | 643 | |||
JX117423.1 | - | 4 | 606 | |||
JX117424.1 | - | 4 | 606 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000629145.1 | - | 505 | Chr. - - [-] on Build |