Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPR1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108075.1 | 2 | 605 | |||
GenBank mRNA | AL832729.1 | - | 1 | 2030 | ||
BI523552.1 | - | 2 | 633 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX359013.2 | - | 2 | 582 | Chr. 3 - 4490972 - 4493140 [-] on Build GRCh38 |