Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026932.1 | 1 | 690 | |||
GenBank mRNA | AK093606.1 | - | 1 | 690 | ||
AK097703.1 | - | 1 | 1565 |