Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNRD1ASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF032109.2 | - | 3 | 3247 | ||
AF032110.1 | - | 3 | 1085 | |||
AF032111.1 | - | 1 | 320 | |||
BX648746.1 | - | 3 | 736 |