Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101926934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134642.1 | 2 | 249 | |||
NR_134643.1 | 2 | 247 | ||||
NR_134644.1 | 2 | 174 | ||||
GenBank mRNA | BX114358.1 | - | 2 | 250 |