Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102724009 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120665.1 | 1 | 50 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000438266.1 | - | 1 | 50 | Chr. 17 - 5019214 - 5020093 [-] on Build GRCh38 |