Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105373764 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136171.1 | 4 | 487 | |||
GenBank mRNA | BX094187.1 | - | 4 | 487 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000450227.1 | - | 2 | 187 | Chr. 2 - 177698490 - 177723289 [+] on Build GRCh38 |