Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105373764 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136171.1 | 4 | 438 | |||
GenBank mRNA | BX094187.1 | - | 4 | 438 |