Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A12-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120614.1 | 3 | 487 | |||
GenBank mRNA | BX089497.1 | - | 3 | 468 | ||
DB088249.1 | - | 3 | 487 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000454270.5 | - | 3 | 414 | Chr. 10 - 89456064 - 89468140 [+] on Build GRCh38 |