Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNA1G-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038439.1 | 2 | 417 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY134540.1 | - | 2 | 410 | Chr. 17 - 50557159 - 50562101 [-] on Build GRCh38 |