Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929517 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120581.1 | 2 | 250 | |||
GenBank mRNA | BC043517.1 | - | 2 | 250 | ||
KP742344.2 | - | 2 | 250 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY024299.1 | - | 2 | 248 | Chr. 11 - 128526145 - 128526829 [+] on Build GRCh38 |