Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102723330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132279.1 | 2 | 521 | |||
GenBank mRNA | BX103002.1 | - | 2 | 521 |