Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKCQ-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036502.1 | 3 | 1081 | |||
NR_036503.1 | 2 | 1017 | ||||
GenBank mRNA | AY007155.1 | - | 2 | 903 | ||
DB554966.1 | - | 2 | 327 |