Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101928402 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130770.1 | 1 | 107 | |||
GenBank mRNA | DW416112.1 | - | 1 | 47 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000426367.1 | - | 1 | 107 | Chr. X - 123959233 - 123961341 [-] on Build GRCh38 |