Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102724312 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125994.1 | 1 | 90 | |||
NR_125995.1 | 1 | 90 | ||||
NR_125996.1 | 1 | 90 | ||||
GenBank mRNA | BM699504.1 | - | 1 | 90 | ||
BM726062.1 | - | 1 | 37 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000417917.1 | - | 1 | 37 | Chr. 1 - 1430664 - 1434520 [-] on Build GRCh38 |