Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126363.1 | 2 | 242 | |||
GenBank mRNA | BC038196.1 | - | 2 | 242 | ||
BC038576.1 | - | 2 | 242 |