Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3D21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017002340.1 | XP_016857829.1 | 18 | 3100 | ||
XM_017002341.1 | XP_016857830.1 | 17 | 3094 |
Gene Symbol: LOC100127947 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY240960.1 | - | 2 | 1103 |