Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNHG16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038108.1 | 4 | 541 | |||
NR_038109.1 | 4 | 537 | ||||
NR_038110.1 | 3 | 442 | ||||
NR_038111.1 | 3 | 438 | ||||
GenBank mRNA | AB073602.1 | - | 3 | 427 | ||
AB447886.1 | - | 4 | 536 | |||
AK055935.1 | - | 4 | 495 | |||
AK129904.1 | - | 3 | 434 | |||
BC042949.2 | - | 3 | 1610 | |||
BC100293.1 | - | 4 | 469 | |||
BC112893.1 | - | 1 | 240 | |||
BX647993.1 | - | 3 | 397 |